Automated microbial detection systems for water quality, TECTA™
Supplier: PATHOGEN DETECTION SYSTEMS
TECTA-CCA-48 TECTAB16A230
PDSYTECTA-CCA-48EA 0 SGD
PDSYTECTA-CCA-48 PDSYTECTAB16A230
Automated microbial detection systems for water quality, TECTA™
Monitoring Systems Water Testing Monitoring Systems
The TECTA™ B16 and TECTA™ B4 are complete, self-contained automated microbiology testing systems for water quality capable of producing laboratory grade results with unprecedented 'time-to-result' performance. Tests can be inititated at any time, with automatic reporting of results that are sent directly via email notification to any device immediately upon detection.
  • Rapid alerting with no fixed incubation delay
  • High reliability with minimal handling of samples
  • No sample preparation or visual interpretation of results required
  • High dynamic range
More Product Information