Sampling swabs
Supplier: PURITAN MEDICAL
25-806-1PD
PMPC25-806-1PDEA 0 SGD
PMPC25-806-1PD
Sampling swabs
Swabs Swabs for Sampling
This synthetic tipped applicator is made of medical-grade polyester fibre that is carded in our factory to maintain consistency.
  • Standard tip with medical grade quality
  • Non-absorbent no fatty acids or latent sugars for specimen transport, lab work, and diagnostic kits
  • Bonded securely to a semi-flexible PS handle by an aqueous based adhesive
Applications include diagnostic specimen collection, patient care, cell collection for DNA testing and gynaecological screening.
Packaging: 10 boxes per case of 100 individually wrapped packages.
Order Now
SPECIFICATIONS
Environmentally Preferable