Dry Transport Systems
Supplier: PURITAN MEDICAL
25-8061PCBT
PMPC25-8061PCBTEA 178 SGD
PMPC25-8061PCBT
Dry Transport Systems
Swabs Transport Swabs
Puritan's new dry transport system securely holds a swab in a round bottom tube made from virgin polypropylene resin, creating a safe and sterile environment to accommodate many different styles of collection devices.
  • High quality USP grade cotton tip
  • Specimen collection for testing and screening
  • Cell collection for DNA testing
  • Shelf life is 3 years
The injection molded cap assures a precision fit for multiple handle styles eliminating any foreign adhesive which may interfere with the specimen. Tubes include a distinctive label describing productMore Product Information