CSLAB Interchangeable Syringes
Supplier: CADENCE SCIENCE
2406
POPR2406EA 0 SGD
POPR2406
CSLAB Interchangeable Syringes
Syringes General-Purpose Syringes
These Cadence Science® CSLAB interchangeable syringes are manufactured to ensure the perfect mating of barrels and plungers. Any interchangeable syringe of a particular size will fit any interchangeable barrel of that same size.
  • Precision shrinking ensures improved fit
  • Coloured plunger ends are fused around bottom beveled rim of plunger to facilitate dosage measurements
  • Made from borosilicate glass providing protection against thermal shock
  • Corrosion resistant
  • Graduations permanently fused into glass barrel and reinforced flanges prevent syringe rolling
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable