Sampling cannulae
Supplier: QUALITY LAB ACCESSORIES
CA8015-PV CA8015-VK CA8475-PV CA8475-VK CA8775-PV CA8775-VK CAN013-ST CAN013-ST-5 CAN013-ST-9 CAN015-PH CAN015-SH CAN475-01 CAN475-HR CAN475-PH CAN475-SH CAN500-60 CAN775-01 CAN775-HR CAN775-PH CAN775-SH CAN900-45 CANRET-25 CANS15-PH PRBFSR-ST PRBNSR-ST
QLAACA8015-PVEA 0 SGD
QLAACA8015-PV QLAACA8015-VK QLAACA8475-PV QLAACA8475-VK QLAACA8775-PV QLAACA8775-VK QLAACAN013-ST QLAACAN013-ST-5 QLAACAN013-ST-9 QLAACAN015-PH QLAACAN015-SH QLAACAN475-01 QLAACAN475-HR QLAACAN475-PH QLAACAN475-SH QLAACAN500-60 QLAACAN775-01 QLAACAN775-HR QLAACAN775-PH QLAACAN775-SH QLAACAN900-45 QLAACANRET-25 QLAACANS15-PH QLAAPRBFSR-ST QLAAPRBNSR-ST
Sampling cannulae
Cannulae
Standard cannula are unique, proprietary configurations for most manual and automated sampling applications.
  • Manufactured in both electropolished 316 stainless steel or PEEK
  • Can accommodate all standard tip filters
  • Optional stoppers set the proper sampling position
The USP defines the sampling point as: 'A zone half way between the top of the media and top of the paddle or basket not less than 1 cm from the vessel wall. The sampling position is critical but so too is the type of sampling cannula (also known as a probe) used.'
Order Now
SPECIFICATIONS
Environmentally Preferable