qPCR cycler, Q
Supplier: Quantabio
95900-4C 95915-20
QUNT95900-4CEA 0 SGD
QUNT95900-4C QUNT95915-20
qPCR cycler, Q
PCR Cyclers
Q qPCR cycler uses a patented magnetic induction technology to rapidly heat samples, coupled with fan-forced air for cooling to acquire data in only 25 minutes. Available as four channel models, the robust optical system acquires all channels simultaneously and allows for running the fastest multiplexed assays.
  • Ultra-fast data acquisition - 35 cycles in 25 minutes (cycle times obtained with fast cycling master mixes and short amplicon assay designs targeting cDNA)
  • Unrivalled performance - detects two-fold expression level differences
  • Portable and compact -
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Gradient -