n-Hexane isomers_mixture_method 98,50 HPLC, Rankem®

Supplier: RANKEM
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

H0200 H0202
RANKH0200EA 0 SGD
RANKH0200 RANKH0202
n-Hexane isomers_mixture_method 98,50 HPLC, Rankem®
n-Hexane

High purity solvent providing stable baseline. Improved specifications enabling better performance and clean chromatograms in the relevant elution range.


  • Batch-to-batch consistency
  • Chemically treated for removal of UV-absorbing, fluorescence -sensitive, aromatic impurities
  • Controlled acidity/alkalinity (≤0,0002 meq/g)


Description: A clear, colourless liquid. Filtered through 0,2 µm filters.

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 66...69 °C (1013 hPa)
Melting Pt: –94,3 °C
Density: 0,674 g/cm³ (20 °C)
Flash Pt: –18 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
REACH: 01-2119480412-44
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) ≤ 10
Guarantee Analysis .
Assay(GC)(Sum Of 5 Isomers And Methyl cyclopentane) ≥ 98.50 %
Weight Per ml (at 20°C) 0.66 - 0.67 g
Identity (IR) Conforms
Maximum Limits Of Impurities .
Residue After Evaporation ≤ 0.0005 %
Titrable Acid ≤ 0.0002 meq/g
Titrable Base ≤ 0.0002 meq/g
Water ( By KF) ≤ 0.02 %
Sulphur Compounds. <= 0.005%
Thiophene Passes test
UV Transmittance .
200 nm ≥ 20 %
210 nm ≥ 50 %
220 nm ≥ 80 %
230 nm ≥ 90 %
From 245 nm ≥ 98 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR