n-Hexane 99%+ HPLC, Rankem®

Supplier: RANKEM
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

H0360 H0365
RANKH0360EA 0 SGD
RANKH0360 RANKH0365
n-Hexane 99%+ HPLC, Rankem®
n-Hexane

High purity solvent providing stable baseline. Improved Specifications enabling better performance and clean chromatograms in the relevant elution range.


  • Batch-to-Batch consistency
  • Chemically treated for removal of UV-absorbing, fluorescence -sensitive, aromatic impurities
  • Controlled acidity/alkalinity ( ≤0.0002 meq/g)


Description: A clear, colourless liquid. Filtered through 0.2 micron filters

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –94,3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
REACH: 01-2119480412-44
Merck Index: 13,04712

Order Now

Specification Test Results

Color (APHA) ≤ 10
Guarantee Analysis .
Assay(GC) Min. 99.00 %
Weight Per ml (at 20°C) 0.659 -- 0.660 g
Identity (IR) Conforms
Maximum Limits Of Impurities .
Residue After Evaporation Max. 0.0005 %
Titrable Acid Max. 0.0002 meq/g
Titrable Base Max. 0.0002 meq/g
Water ( By KF) Max. 0.02 %
UV Transmittance .
Wavelength .
210 nm Min. 60 %
220 nm Min. 80 %
230 nm Min. 90 %
From 245 nm Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR