Phenol ≥99% LR, Laboratory Reagent, Rankem®

Supplier: RANKEM
Danger

P0135
RANKP0135EA 0 SGD
RANKP0135
Phenol ≥99% LR, Laboratory Reagent, Rankem®
Phenol

General lab reagent for routine laboratory use.


  • Suitable for routine laboratory usage and classical analysis


Description: Colourless crystalline mass.

Formula: C₆H₅OH
Boiling Pt: 181,8 °C (1013 hPa)
Melting Pt: 40,8 °C
Density: 1,06 g/cm³ (20 °C)
Flash Pt: 81 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1,II

Order Now

Specification Test Results

Guarantee Analysis .
Appearance Of Solution Passes test
Assay(GC) ≥ 99.00 %
Freezing Point 39.0 - 43.0 °C
Identity (IR) Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR