1-Pentanesulphonic acid sodium salt for HPLC, Rankem®

Supplier: RANKEM

P2109
RANKP2109EA 10.6 SGD
RANKP2109
1-Pentanesulphonic acid sodium salt for HPLC, Rankem®
1-Pentanesulphonic acid sodium salt
Formula: C₅H₁₁O₃SNa
MDL Number: MFCD00007541
CAS Number: 22767-49-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddb1c23c8a6fe3c156a3aacb62721e1d1a34efb0&vid=1936dcdb47ba374ce21fd758660f7ca6ba95d254&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR