Water for HPLC, Rankem®

Supplier: RANKEM

Synonyms: Hydrogen oxide

W0020
RANKW0020EA 1.2 SGD
RANKW0020
Water for HPLC, Rankem®
Water

High purity solvent providing stable baseline.


  • Batch-to-Batch consistency
  • Controlled NVM
  • Optimised packaging


Description: A clear, colourless liquid, filtered through 0,2 µm filters.

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2

Order Now

Specification Test Results

Guarantee Analysis .
Non volatile matter Max. 0.0003 %
UV Absorbance .
Wavelength .
210 nm Max. 0.004
254 nm Max. 0.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR