Water LR, Rankem®

Supplier: RANKEM

Synonyms: Hydrogen oxide

W0030
RANKW0030EA 5.5 SGD
RANKW0030
Water LR, Rankem®
Water

General lab reagent Suitable for routine laboratory usage and classical analysis.


  • A clear, colourless liquid
  • Controlled TDS

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2

Order Now

Specification Test Results

Guarantee Analysis .
pH 5.00 - 7.50
Maximum Limits Of Impurities .
Chloride (Cl) Passes test
Sulphate (SO₄) Passes test
Iron (Fe) Passes test
Heavy Metal (As Pb) Passes test
Nitrate (NO₃) Passes test
Ammonium (NH₄) Passes test
Calcium Magnesium Passes test
Total Dissolved Solid Max. 0.01%
Oxidisable Substances Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR