Transport trolley, folding
Supplier: RAUTEC
40002
RAUT40002EA 0 SGD
RAUT40002
Transport trolley, folding
Trolleys
Easily manoeuvrable, multi-purpose, two-level trolley. Can be folded flat in seconds at the press of a button. Equipped with one folding box with 46 litre capacity (additional folding boxes can be ordered by quoting Cat. No. RAUT2050049).
  • Steerable castors for excellent manoeuvrability, parking brake on the rear wheels
  • Each tray can carry one folding box at the same time
  • Load capacity: bottom level 40 kg, top level 20 kg
  • W×D×H: Folded - 710×550×190 mm; assembled - 900×580×1020 mm
Aluminium frame with plastic construction.
Order Now
SPECIFICATIONS
Environmentally Preferable