Acetic acid 12% Reag. Ph. Eur. 1000402

Supplier: REAGECON
Warning

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

1000402
REGE1000402EA 0 SGD
REGE1000402
Acetic acid 12% Reag. Ph. Eur. 1000402
Acetic acid
Formula: H₃CCOOH
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2790
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR