Ammonia solution 10% Reag. Ph. Eur. 1004702

Supplier: REAGECON
Danger

1004702
REGE1004702EA 0 SGD
REGE1004702
Ammonia solution 10% Reag. Ph. Eur. 1004702
Ammonia solution
Formula: NH₃
Density: 0,92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR