Conductivity standards, KCl, referenced at 25 °C
Supplier: REAGECON
CSKC10 CSKC100 CSKC1000 CSKC100M CSKC10M CSKC12880 CSKC150M CSKC20 CSKC200 CSKC200M CSKC20M CSKC300M CSKC3M CSKC50M CSKC5M CSKC84 CSKCL CSKC136 CSKC350M CSKC13 CSKC2M CSKC238 CSKC300 CSKC400 CSKC111800 CSKC2500 CSKC25 CSKC185 CSKC250 CSKC30M CSKC400M CSKC40M CSKC1025
REGECSKC10EA 210 SGD
REGECSKC10 REGECSKC100 REGECSKC1000 REGECSKC100M REGECSKC10M REGECSKC12880 REGECSKC150M REGECSKC20 REGECSKC200 REGECSKC200M REGECSKC20M REGECSKC300M REGECSKC3M REGECSKC50M REGECSKC5M REGECSKC84 REGECSKCL REGECSKC136 REGECSKC350M REGECSKC13 REGECSKC2M REGECSKC238 REGECSKC300 REGECSKC400 REGECSKC111800 REGECSKC2500 REGECSKC25 REGECSKC185 REGECSKC250 REGECSKC30M REGECSKC400M REGECSKC40M REGECSKC1025
Conductivity standards, KCl, referenced at 25 °C
Standards Conductivity Standards
These conductivity standards are aqueous and so avoid the problems of matrix errors and high temperature dependency of nonaqueous conductivity standards.
  • With an accuracy of ±1,0% (except 1,3 µS)
  • Temperature dependence data, lot no. and expiry date are printed on product labels
These solutions standardised at 25 °C are directly traceable to N.I.S.T standard reference materials and are tested using an ISO 17025 accredited method.
Order Now
SPECIFICATIONS
Environmentally Preferable