Hydrochloric acid 10%

Supplier: REAGECON
Warning

H10WV1
REGEH10WV1EA 0 SGD
REGEH10WV1
Hydrochloric acid 10%
Hydrochloric acid
Formula: HCl
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR