Redox standards
Supplier: REAGECON
RS124 RS200 RS300 RS465 RS650 RS250 RS600
REGERS124EA 0 SGD
REGERS124 REGERS200 REGERS300 REGERS465 REGERS650 REGERS250 REGERS600
Redox standards
Standards Redox/ORP Standards
The potentials quoted are of platinum electrode with reference to Ag/AgCl (3 M KCl).
Order Now
SPECIFICATIONS
Environmentally Preferable