Blood Bank Refrigerators
Supplier: REVCO TECHNOLOGIES
REB2304V REB404V REB5004V REB1204V
REVCREB2304VEA 0 SGD
REVCREB2304V REVCREB404V REVCREB5004V REVCREB1204V
Blood Bank Refrigerators
Refrigerators
These blood bank refrigerators store whole blood and blood components.
  • Second cabinet temperature probe
  • Keyed on/off switch with setpoint security
  • Built-in chart recorder and remote alarm contacts
  • Heat-free defrost for maximum temperature uniformity
  • Scratch-resistant painted chamber and cabinet material
  • Bright LED interior lighting and optional 4 to 20 mA output
The features include microprocessor control systems, positive airflow systems, industrial-quality cabinet construction with adjustable levelling feet and extra-strength refrigerationMore Product Information