Pharmacy refrigerators
Supplier: REVCO TECHNOLOGIES
RPR5004V
REVCRPR5004VEA 0 SGD
REVCRPR5004V
Pharmacy refrigerators
Refrigerators
These Pharmacy refrigerators delivers stable temperature control over a range of +1°C to +8°C. It is configured easily for accurate storage and retrieval of pharmacy products.
  • Keyed on/off switch with setpoint security
  • Advanced defrost sensor manages the defrost cycle, minimizing frost buildup
  • Feature full extension stainless-steel roll-out drawers
  • Spring-loaded, self closing doors with 90° stay-open feature
  • High-density, CFC-free urethane foam insulation protects cabinet from ambient temperature fluctuations
  • Simplified operation controls with enhanced
More Product Information