Cresol red sodium salt 0,04% in aqueous solution

Supplier: Ricca Chemical

2450-16
RICC2450-16EA 0 SGD
RICC2450-16
Cresol red sodium salt 0,04% in aqueous solution
Cresol red sodium salt
Formula: C₂₁H₁₇NaO₅S
MDL Number: MFCD00001618
CAS Number: 62625-29-0
EINECS: 263-654-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR