Hydrochloric acid 0.05 mol/l (0.05 N)

Supplier: Ricca Chemical

Synonyms: Hydrochloric acid solution, Muriatic acid

3597-1
RICC3597-1EA 0 SGD
RICC3597-1
Hydrochloric acid 0.05 mol/l (0.05 N)
Hydrochloric acid
Formula: HCl
Density: 1,004 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR