Methyl red 0,1% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical

Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid

5060-32
RICC5060-32EA 0 SGD
RICC5060-32
Methyl red 0,1% (w/v) in aqueous solution indicator
Methyl red
Formula: C₁₅H₁₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00002425
CAS Number: 493-52-7
EINECS: 212-682-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR