Potassium nitrate 100 g/l in aqueous solution

Supplier: Ricca Chemical

Synonyms: Nitrate of Potash

6317-16
RICC6317-16EA 0 SGD
RICC6317-16
Potassium nitrate 100 g/l in aqueous solution
Potassium nitrate
Formula: KNO₃
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR