Starch 1% (w/v) in aqueous solution for iodometric titrations mercury-free

Supplier: Ricca Chemical

8050-32
RICC8050-32EA 0 SGD
RICC8050-32
Starch 1% (w/v) in aqueous solution for iodometric titrations mercury-free
Starch

Indicator. Translucent liquid. For general iodometric titrations.


  • Preserved with 0.01–0.2% salicylic acid
  • Mercury free.

Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR