Thymolphthalein 0,5 g/l in ethanol 90% (v/v)

Supplier: Ricca Chemical
Danger

8450-16
RICC8450-16EA 0 SGD
RICC8450-16
Thymolphthalein 0,5 g/l in ethanol 90% (v/v)
Thymolphthaleine
Formula: C₂₈H₃₀O₄
Storage Temperature: Ambient
MDL Number: MFCD00005909
CAS Number: 125-20-2
EINECS: 204-729-7
UN: 1170
ADR: 3,II
Merck Index: 13,09478

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR