Ammonium heptamolybdate USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R0674000-120B
RICCR0674000-120BEA 0 SGD
RICCR0674000-120B
Ammonium heptamolybdate USP Test Solution (TS)
Ammonium heptamolybdate solutions
Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Boiling Pt: ~ 100 °C
Density: 1,7000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
UN: 3264
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear to hazy, colorless to light green with slight nitric acid odor
Suitability for Use Yellow precipitate with Phosphate

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR