EDTA disodium salt 0.1 mol/l in aqueous solution titrant

Supplier: Ricca Chemical

R2730000-1A
RICCR2730000-1AEA 0 SGD
RICCR2730000-1A
EDTA disodium salt 0.1 mol/l in aqueous solution titrant
EDTA disodium salt
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 200-449-4
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR