Mercury(II) acetate 6% (w/v) in glacial acetic acid USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

Synonyms: Mercury diacetate, Mercuric acetate, Acetic acid mercury (II) salt

R4629000-1C
RICCR4629000-1CEA 0 SGD
RICCR4629000-1C
Mercury(II) acetate 6% (w/v) in glacial acetic acid USP Test Solution (TS)
Mercury (II) acetate
Formula: (H₃CCOO)₂Hg
Melting Pt: 178…180 °C
Density: 3,27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012165
CAS Number: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1,II
Merck Index: 13,05898

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR