Spatula set
Supplier: RSG
000.105
RSGA000.105EA 87.8 SGD
RSGA000.105
Spatula set
Spatulas
Stainless steel 18/10. Useful combination of double spatulas, spoon spatulas and forceps. Universal usage in laboratories and industry.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Comes in practical imitation leather case
Each set includes a 150×5 and 150×7 mm spoon spatula, a 150×9 mm double spatula, a 150×4 mm micro double spatula, a 150×9 mm powder spatula, a 150×4 mm micro powder spatula, as well as tweezers (straight, blunt, 145 mm long).
Each set includes a 150×5 and 150×7 mmMore Product Information