Forceps set
Supplier: RSG
000.120
RSGA000.120EA 100 SGD
RSGA000.120
Forceps set
Forceps
Set contains four pairs of forceps made from stainless steel:
1× fine pointed
1× blunt ended
1× fine with guide pin
1× cover slip
Guidebook and case