Laboratory spoon
Supplier: RSG
026.200
RSGA026.200EA 14.6 SGD
RSGA026.200
Laboratory spoon
Spoons
Stainless steel 18/10, anti-magnetic. Multipurpose spoon for a wide range of uses.
  • Open ended spoon, with flat handle
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Order Now
SPECIFICATIONS
Environmentally Preferable