Pharmacist's spoon
Supplier: RSG
028.150
RSGA028.150EA 0 SGD
RSGA028.150
Pharmacist's spoon
Spoons
Stainless steel 18/10, anti-magnetic. Multipurpose spoon for a wide range of uses.
  • Narrow shape with extra long, flat handle
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean