Spoon, powder
Supplier: RSG
029.170
RSGA029.170EA 0 SGD
RSGA029.170
Spoon, powder
Spoons
Stainless steel 18/10. Multipurpose spoon for a wide range of uses.
  • With cylindrical handle
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean