Dissection set
Supplier: RSG
066.906
RSGA066.906EA 117 SGD
RSGA066.906
Dissection set
Dissection Sets
Dissecting set with six parts made from stainless steel, packed in a pouch.
  • 1 spoon spatula, 150 mm
  • 1 pointed tweezer, 130 mm
  • 1 powder spatula, 150/9 mm
  • 1 scalpel, 125 mm
  • 1 scissors straight, 100 mm
  • 1 spatula with blade, 150 mm