Spatulas, micro, set
Supplier: RSG
087.000
RSGA087.000EA 0 SGD
RSGA087.000
Spatulas, micro, set
Spatulas
Stainless steel. Set of four 150 mm (overall length) micro spatulas with black plastic handle.
  • Choice of 4 blade widths (3, 4, 5, 6 mm) in one set
  • 45 mm blade length