Micro powder spatula set
Supplier: RSG
087.300
RSGA087.300EA 0 SGD
RSGA087.300
Micro powder spatula set
Spatulas
Stainless steel 18/10 blade with black plastic handle.
  • Four-piece set, each with a 3, 4, 5 and 6 mm wide powder spatula
  • Blade has excellent chemical and thermal resistance
  • Blades are semicircular and angled