Forceps
Supplier: RSG
111.010 113.010
RSGA111.010EA 0 SGD
RSGA111.010 RSGA113.010
Forceps
Forceps
Steel mainspring, lightweight. Special forceps for particularly accurate and delicate work.
  • Lightweight
  • Available in two designs
  • Extremely flexible