Support rings
Supplier: RSG
280.070
RSGA280.070EA 0 SGD
RSGA280.070
Support rings
Retort Rings
These heavy-duty support rings are made from stainless steel.
  • 8 mm rod
  • Available with rubber covered rings