Micro powder spoon
Supplier: RSG
533.005
RSGA533.005EA 0 SGD
RSGA533.005
Micro powder spoon
Spoons
Stainless steel 18/10, anti-magnetic. With hexagonal handle for easier handling.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean