Spoon spatulas, micro
Supplier: RSG
985.130
RSGA985.130EA 5 SGD
RSGA985.130
Spoon spatulas, micro
Spatulas
Stainless steel 18/10, anti-magnetic.
  • With round handle and one bent spoon
  • Excellent temperature resistance, autoclavable
  • Good resistance to chemicals
  • Very flexible