BRUTE® Dollies
Supplier: Rubbermaid Commercial Products
FG265000BLA
RUBBFG265000BLAEA 0 SGD
RUBBFG265000BLA
BRUTE® Dollies
Trolleys
High-performance dollies for efficient waste management and material transport.
  • Easy mobility
  • High load capacity
  • Durable construction
The BRUTE® dolly (129-4204P) is designed to smoothly and efficiently transport 76, 121, 167, and 208 L BRUTE® containers with ease. Easy mobility: Provides easy mobility and maneuverability when collecting and transporting heavy loads. High load capacity: Supports loads up to 113 kg (250 lbs.). Secure Design: Innovative twist-lock design keeps containers secure. Quiet Operation: five non-marking casters move quietly andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable