Spatula, 'V' shape
Supplier: Sampling Systems
1095S-150
SAMP1095S-150EA 0 SGD
SAMP1095S-150
Spatula, 'V' shape
Spatulas
This V spatula made from stainless steel 316 is ideal for surface sampling of powders and core sampling of viscous material like cheese, pastes and gels.
  • Polished finish
  • Fully welded, crevice free construction
  • ‘V’ shape allows easy discharge of the powders into containers or bags
Order Now
SPECIFICATIONS
Environmentally Preferable