TeleScoop Water Samplers
Supplier: Sampling Systems
5065A-1001 5065A-1201 5065A-2501
SAMP5065A-1001EA 0 SGD
SAMP5065A-1001 SAMP5065A-1201 SAMP5065A-2501
TeleScoop Water Samplers
Samplers Fluid Samplers
TeleScoop water sampler is the ultimate extendable water sampler for liquid and water sampling.
  • Range of telescopic poles upto 6 m in length
  • Robust and lightweight design
  • Multi angle elbow design
  • Quick and easily interchangeable accessories - no tools needed
  • Variety of compatible sampling accessories
  • Ideal for sampling from river banks or overhead gantries
The TeleScoop water sampler is the latest extendable sampler for sampling waters and liquids from hard-to-reach places. The extendable rods reach upto 6 m in length and the robust yet lightweightMore Product Information