Hand paddle with holes, Sampling Systems
Supplier: Sampling Systems
8026P-540
SAMP8026P-540EA 0 SGD
SAMP8026P-540
Hand paddle with holes, Sampling Systems
Stirrers Stirring Rods
The hand paddle with holes is perfect for stirring and mixing liquids.
  • Short, ergonomic handle for ease of use
  • Holes allow for easy mixing
  • Mde from robust chemicall resistant PP
  • 133 mm blade width
The hand paddle is excellent for stirring liquids in large containers, and the ergonomic handle allows it to be used with one hand or with both hands.