Liquid samplers, SteriWare®
Supplier: Sampling Systems
8051H-450 8051H-450S 8051H-900 8051H-900S 8051H-1100 8051H-1100S
SAMP8051H-450EA 0 SGD
SAMP8051H-450 SAMP8051H-450S SAMP8051H-900 SAMP8051H-900S SAMP8051H-1100 SAMP8051H-1100S
Liquid samplers, SteriWare®
Samplers Fluid Samplers
These disposable Jumbo pipettes are ideal for taking samples from low viscosity liquids. Made from 100% virgin HDPE.
  • Quick and easy to use
  • Colour-coded tip indicates which end to open and hold
  • Cross-contamination risk is eliminated - can be used straight from the bag
  • No cleaning is required before or after sampling
  • Available as gamma irradiated sterile (SAL 10⁻⁶)
  • Suitable for use in ATEX zones 0, 1 and 2 (450 and 900 mm lengths only)
Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc).
Certifications: FDAMore Product Information