Quick Clip
Supplier: Sampling Systems
8219A-220
SAMP8219A-220EA 0 SGD
SAMP8219A-220
Quick Clip
Clamps General Purpose Clamps
Quick clip provides an instant seal for bags.
  • Provides an instant powder- and water-tight seal for bags
Quick clips can be used over and over again, providing a convenient and cost-eefective solution to sealing different types of bags.