Re-Usable Cleaning Coupons
Supplier: Sampling Systems
8644SIL-100
SAMP8644SIL-100EA 0 SGD
SAMP8644SIL-100
Re-Usable Cleaning Coupons
Wipes Cleaning Kits and Pads
Cleaning coupons are great for validating a cleaning regime.
  • All popular sizes available
  • Wide variety of materials and finishes
Sampling Systems' extensive range of cleaning coupons come in a diverse selection of materials, sizes, and finishes. They are ideal for validating cleaning regimes. Popular materials include 316 stainless steel, PTFE, aluminium, and HDPE.