Micro spatula, stainless steel
Supplier: Sampling Systems
A100-20
SAMPA100-20EA 0 SGD
SAMPA100-20
Micro spatula, stainless steel
Spatulas
Perfect for transferring small amounts of powder.
  • Ideal for transferring powders, granules and cream
  • Used for surface sampling and dispensing
  • Crevice free
The range of stainless steel microspatulas are perfect for transferring very small amounts of powders, creams and granules and are ideal for use in pharma, food and chemical companies. The crevice free stainless steel construction means the product can be easily cleaned between uses.
Order Now
SPECIFICATIONS
Environmentally Preferable