Economy scoop
Supplier: Sampling Systems
A304-450
SAMPA304-450EA 0 SGD
SAMPA304-450
Economy scoop
Scoops
These scoops made from 304 stainless steel are ideal for general industrial use.
  • High polish
  • Light duty construction
  • Round handle
Please note: This scoop is not suitable for GMP applications.